missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 30% (w/v) Electrolyte Solution, Spectrum™ Chemical

Catalog No. 18612708
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
55 gal.
Packaging:
Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612708 500 mL Poly Bottle
18612709 1 L Poly Bottle
18612710 4 L Poly Bottle
18612711 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612708 Supplier Spectrum Chemical Mfg Cor Supplier No. P385500MLPL
Only null left

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5

Specifications

Quantity 500 mL
Percent Purity 30% ±0.5%
Grade Chemical Solution
Packaging Poly Bottle
Chemical Name or Material Potassium Phosphate Dibasic, Trihydrate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.