missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 30% (w/v), Ricca Chemical
$175.44 - $820.91
Chemical Identifiers
| CAS | 1310-58-3 |
|---|---|
| Molecular Formula | HKO |
| Molecular Weight (g/mol) | 56.11 |
| MDL Number | MFCD00003553 |
| InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
| Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
| PubChem CID | 14797 |
| ChEBI | CHEBI:32035 |
| IUPAC Name | potassium hydroxide |
| SMILES | [OH-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6153-16
|
Ricca Chemical Company
615316 |
500 mL | Natural Poly Bottle |
N/A
|
|
||||
|
6153-32
|
Ricca Chemical Company
615332 |
1 L | Natural Poly Bottle |
N/A
|
|
||||
|
6153-1
|
Ricca Chemical Company
61531 |
4 L | Natural Poly Bottle |
N/A
|
|
||||
|
61532-5
|
Ricca Chemical Company
615325 |
10 L | Cubitainer |
N/A
|
|
||||
Chemical Identifiers
| 1310-58-3 | |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| 14797 | |
| potassium hydroxide |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| CHEBI:32035 | |
| [OH-].[K+] |
Specifications
| 1310-58-3,7732-18-5 | |
| 78.0,23.55 | |
| MFCD00003553 | |
| 500 mL | |
| [OH-].[K+] | |
| 56.11 | |
| CHEBI:32035 | |
| Laboratory | |
| Natural Poly Bottle |
| 75.0,22.63 | |
| HKO | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium hydroxide | |
| 14797 | |
| 29.94 to 30.06% | |
| 30% (w/v) Aqueous Solution | |
| Potassium Hydroxide |