missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 30% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company 615332

 View more versions of this product

Catalog No. 6153-32


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Hydroxide
75.0,22.63
HKO
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
29.94 to 30.06%
30% (w/v) Aqueous Solution
1310-58-3,7732-18-5
78.0,23.55
MFCD00003553
1 L
[OH-].[K+]
56.11
CHEBI:32035
Laboratory
Natural Poly Bottle

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions