missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 10% (w/v), Solution, Spectrum™ Chemical

Catalog No. p-7166159
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612702 500 mL Poly Bottle
18612735 1 L Poly Bottle
18612736 4 L Poly Bottle
18612703 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612702 Supplier Spectrum Chemical Mfg Cor Supplier No. P377500MLPL
Only null left

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5

Specifications

Quantity 500 mL
Percent Purity 10% ±0.3%
Grade Chemical Solution
Packaging Poly Bottle
Chemical Name or Material Potassium Hydroxide Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.