missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 10% (w/v), Solution, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P3771LT

 View more versions of this product

Catalog No. 18612735


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Hydroxide Solution
10% ± 0.3%
10% ±0.3%
Poly Bottle
1310-58-3,7732-18-5
1 L
Chemical Solution

Chemical Identifiers

1310-58-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.