missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 1.0 N Aqueous Solution, Spectrum™ Chemical

Catalog No. 18612697
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612697 500 mL Poly Bottle
18612698 1 L Poly Bottle
18612699 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612697 Supplier Spectrum Chemical Mfg Cor Supplier No. P360500ML
Only null left

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5

Specifications

Concentration 1.0 N ± 0.03 N
Quantity 500 mL
Grade Chemical Solution
Packaging Poly Bottle
Chemical Name or Material Potassium Hydroxide Solution

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.