missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide, 0.5N Standardized Solution in methanol
Description
Potassium hydroxide, 0.5N standardized solution in methanol is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| UN Number | UN3286 |
| Concentration | 0.5N in methanol |
| Solubility Information | Miscible with water. |
| IUPAC Name | potassium;hydroxide |
| Formula Weight | 56.11 |
| Odor | Like alcohol |
| Quantity | 20 L |
| Physical Form | Liquid |
| Chemical Name or Material | Potassium hydroxide Standardized Solution |
Safety and Handling
| Hazard Category | H225-H301+H311+H331-H315-H318-H370 |
| Hazard Statement | GHS H Statement H225-H311-H331-H370-H314 Highly flammable liquid and vapor. Toxic in contact with skin. Toxic if inhaled. Causes damage to organs. Causes severe skin burns and eye damage. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P307+P311-P310-P311-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S. |
| EINECSNumber | 215-181-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only