missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.500 N (N/2) in Isopropyl Alcohol, Ricca Chemical

Catalog No. 6257532
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6257532 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6257532 Supplier Ricca Chemical Company Supplier No. 6257.532
Only null left

Chemical Identifiers

CAS 67-63-0 , 1310-58-3 , 107-15-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Min % 3.89, 94.0, 0.03
CAS Max % 4.05, 98.0, 0.03
Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.500 Normal (N/2) in Isopropyl Alcohol
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide
ShelfLife 12 months