missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 0.250 N (N/4), Ricca Chemical
Supplier: Ricca Chemical Company 622032
Specifications
| Potassium Hydroxide | |
| 1310-58-3,7732-18-5 | |
| 3.09,99.0 | |
| MFCD00003553 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium hydroxide | |
| 14797 | |
| Laboratory | |
| >13 | |
| Liquid |
| Colorless | |
| 2.97,95.0 | |
| HKO | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| [OH-].[K+] | |
| 56.11 | |
| CHEBI:32035 | |
| Natural Poly Bottle | |
| 1 L | |
| 0.250Normal |
Chemical Identifiers
| 1310-58-3 | |
| 56.11 | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| 14797 | |
| potassium hydroxide |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
| CHEBI:32035 | |
| [OH-].[K+] |