missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.100 N (N/10) in Denatured Alcohol, Ricca Chemical

Catalog No. p-3340751
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
Catalog No. Quantity Packaging
6264532 1 L Natural Poly Bottle
626451 4 L Natural Poly Bottle
2 options

Catalog No. 6264532

Supplier: Ricca Chemical Company 6264.532

Only null left

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Min % 1.59, 96.0, 0.03
CAS Max % 1.65, 100.0, 0.03
Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.100 Normal (N/10) in Denatured Alcohol
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide
ShelfLife 12 months