missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.0100 N (N/100) in Methanol, Ricca Chemical

Catalog No. R62640001A
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R62640001A 1 L Natural Poly Bottle
R6264000500 500 mL Natural Poly Bottle
R62640004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R62640001A Supplier Ricca Chemical Company Supplier No. R62640001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67-56-1 | HKO | 56.11 g/mol

Chemical Identifiers

CAS 67-56-1 , 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

pH 12
Color Colorless
Boiling Point ∼64.5°C
Melting Point -98°C
CAS Min % 0.07, 99.92, 0.01
CAS Max % 0.07, 99.92, 0.01
Density 0.8
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, columbian spirit
Solubility Information Miscible at 20°C
Quantity 1 L
Flash Point 12°C
Grade Laboratory
Odor Slight, Alcohol-Like
Packaging Natural Poly Bottle
Vapor Density 1.1
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.0100 Normal (N/100) in Methanol
Show More Show Less
ShelfLife 12 months
Recommended Storage Ambient Flammable