missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroquinonesulfonate 98.0+%, TCI America™
$53.90 - $211.77
Chemical Identifiers
| CAS | 21799-87-1 |
|---|---|
| Molecular Formula | C6H5KO5S |
| Molecular Weight (g/mol) | 228.259 |
| MDL Number | MFCD00007475 |
| InChI Key | VKDSBABHIXQFKH-UHFFFAOYSA-M |
| PubChem CID | 23672329 |
| IUPAC Name | potassium;2,5-dihydroxybenzenesulfonate |
| SMILES | C1=CC(=C(C=C1O)S(=O)(=O)[O-])O.[K+] |
Chemical Identifiers
| 21799-87-1 | |
| 228.259 | |
| VKDSBABHIXQFKH-UHFFFAOYSA-M | |
| potassium;2,5-dihydroxybenzenesulfonate |
| C6H5KO5S | |
| MFCD00007475 | |
| 23672329 | |
| C1=CC(=C(C=C1O)S(=O)(=O)[O-])O.[K+] |
Specifications
| 21799-87-1 | |
| C6H5KO5S | |
| 25 g | |
| VKDSBABHIXQFKH-UHFFFAOYSA-M | |
| potassium;2,5-dihydroxybenzenesulfonate | |
| 23672329 | |
| ≥98.0% (T) | |
| Potassium Hydroquinonesulfonate |
| White-Yellow | |
| MFCD00007475 | |
| potassium 2,5-dihydroxybenzenesulfonate, hydroquinonesulfonic acid potassium salt, potassium2,5-dihydroxybenzenesulfonate, unii-249e3f00ep, benzenesulfonic acid, 2,5-dihydroxy-, monopotassium salt, 2,5-dihydroxybenzenesulfonic acid potassium salt, potassium hydroquinonesulfonate, hydroquinone sulfonic acid potassium salt, hydroquinone sulfonic acid, potassium salt, benzenesulfonic acid, 2,5-dihydroxy-, potassium salt 1:1 | |
| C1=CC(=C(C=C1O)S(=O)(=O)[O-])O.[K+] | |
| 228.259 | |
| 228.26 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2017
TSCA : Yes