missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroquinonesulfonate 98.0+%, TCI America™

Catalog No. H059325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H059325G 25 g
H0593500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H059325G Supplier TCI America Supplier No. H059325G

Chemical Identifiers

CAS 21799-87-1
Molecular Formula C6H5KO5S
Molecular Weight (g/mol) 228.259
MDL Number MFCD00007475
InChI Key VKDSBABHIXQFKH-UHFFFAOYSA-M
PubChem CID 23672329
IUPAC Name potassium;2,5-dihydroxybenzenesulfonate
SMILES C1=CC(=C(C=C1O)S(=O)(=O)[O-])O.[K+]

Specifications

Color White-Yellow
Quantity 25 g
Synonym potassium 2,5-dihydroxybenzenesulfonate, hydroquinonesulfonic acid potassium salt, potassium2,5-dihydroxybenzenesulfonate, unii-249e3f00ep, benzenesulfonic acid, 2,5-dihydroxy-, monopotassium salt, 2,5-dihydroxybenzenesulfonic acid potassium salt, potassium hydroquinonesulfonate, hydroquinone sulfonic acid potassium salt, hydroquinone sulfonic acid, potassium salt, benzenesulfonic acid, 2,5-dihydroxy-, potassium salt 1:1
Formula Weight 228.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Hydroquinonesulfonate
EINECSNumber (3)-2017
TSCA Yes