missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen phthalate, 0.05N Standardized Solution

Catalog No. p-7050104
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35603AP 500 mL
AA35603K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35603AP Supplier Thermo Scientific Chemicals Supplier No. 035603AP
Only null left

CAS: 877-24-7 | C8H5KO4 | 204.222 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
MDL Number MFCD00013070
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Concentration 0.05N
Formula Weight 204.23
Odor Odorless
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Potassium hydrogen phthalate, Standardized Solution
EINECSNumber 212-889-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only