missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen phosphate trihydrate, Reagent Grade

Catalog No. AA3336530
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3336530 250 g
AA33365A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3336530 Supplier Thermo Scientific Chemicals Supplier No. 03336530

CAS: 16788-57-1 | H7K2O7P | 228.22 g/mol

Potassium hydrogen phosphate trihydrate is widely used to prepare buffer solutions in molecular biology, biochemistry and chromatography. It is involved in the production of trypticase soy agar and then utilized for culturing bacteria. As a food additive, it is used in dry powder beverages, mineral supplements and imitation dairy creamers. It is also used as a stabilizer for non-dairy coffee creamers. Further, it is a common source of phosphorous and potassium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16788-57-1
Molecular Formula H7K2O7P
Molecular Weight (g/mol) 228.22
MDL Number MFCD00149926
InChI Key XQGPKZUNMMFTAL-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate
PubChem CID 16217523
IUPAC Name dipotassium;hydrogen phosphate;trihydrate
SMILES O.O.O.[K+].[K+].OP([O-])([O-])=O

Specifications

Physical Form Crystalline
Quantity 250 g
Merck Index 14,7658
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 228.23 (174.18 Anhydrous)
Grade Reagent
Sensitivity Hygroscopic
Chemical Name or Material Potassium hydrogen phosphate trihydrate
EINECSNumber 231-834-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only