Learn More
Potassium hydrogen L-tartrate, 98+%
CAS: 868-14-4 | C4H5KO6 | 188.18 g/mol
Supplier: Thermo Scientific Chemicals A152810B
Description
Potassium hydrogen L-tartrate is used for stabilizing egg whites, whipped cream and for anti-caking and thickening. It is utilized for the prevention of crystallization of sugar syrups and reduces the discoloration of boiled vegetables. It is an active component in baking powder. It is used in household items in association with lemon juice or white vinegar to clean metals such as brass, aluminum and copper. Furthermore, it serves as a primary reference standard for a pH buffer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium hydrogen L-tartrate is used for stabilizing egg whites, whipped cream and for anti-caking and thickening. It is utilized for the prevention of crystallization of sugar syrups and reduces the discoloration of boiled vegetables. It is an active component in baking powder. It is used in household items in association with lemon juice or white vinegar to clean metals such as brass, aluminum and copper. Furthermore, it serves as a primary reference standard for a pH buffer.
Solubility
Soluble in water and dilute mineral acid. Insoluble in alcohol.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Potassium hydrogen L-tartrate | |
| ∼267°C (decomposition) | |
| C4H5KO6 | |
| 1000 g | |
| 14,7615 | |
| Soluble in water and dilute mineral acid. Insoluble in alcohol. | |
| [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O | |
| 188.18 | |
| 188.18 |
| 868-14-4 | |
| 1.954 | |
| MFCD00065392,MFCD00071626,MFCD00064206 | |
| 6119985 | |
| potassium bitartrate, l +-potassium hydrogen tartrate, potassium 3-carboxy-2,3-dihydroxypropanoate, monopotassium tartrate, potassium acid tartrate, kaliumtartrat, potassium hydrogentartrate, 1-potassium hydrogentartrate, ksc657e1t, butanedioic acid, 2,3-dihydroxy-2r,3r-, potassium salt 1:1 | |
| KYKNRZGSIGMXFH-ZVGUSBNCSA-M | |
| 2,3-dihydroxybutanedioic acid;potassium | |
| 24193652 | |
| ≥98% |
Chemical Identifiers
| 868-14-4 | |
| 188.18 | |
| KYKNRZGSIGMXFH-ZVGUSBNCSA-M | |
| 2,3-dihydroxybutanedioic acid;potassium |
| C4H5KO6 | |
| MFCD00065392,MFCD00071626,MFCD00064206 | |
| potassium bitartrate, l +-potassium hydrogen tartrate, potassium 3-carboxy-2,3-dihydroxypropanoate, monopotassium tartrate, potassium acid tartrate, kaliumtartrat, potassium hydrogentartrate, 1-potassium hydrogentartrate, ksc657e1t, butanedioic acid, 2,3-dihydroxy-2r,3r-, potassium salt 1:1 | |
| [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O |
Safety and Handling
EINECSNumber : 212-769-1
RTECSNumber : WW8223000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only