missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen iodate, 0.025N Standardized Solution

Catalog No. AA35600K2
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35600K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA35600K2 Supplier Thermo Scientific Chemicals Supplier No. 035600K2

CAS: 13455-24-8 | HI2KO6 | 389.909 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13455-24-8
Molecular Formula HI2KO6
Molecular Weight (g/mol) 389.909
MDL Number MFCD00011400
InChI Key ACAYDTMSDROWHW-UHFFFAOYSA-M
Synonym potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a
PubChem CID 23700942
IUPAC Name potassium;iodic acid;iodate
SMILES OI(=O)=O.[O-]I(=O)=O.[K+]

Specifications

Concentration 0.025N
Formula Weight 389.91
Odor Odorless
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Potassium hydrogen iodate, Standardized Solution
EINECSNumber 236-650-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only