missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen fluoride, 99+%, extra pure, Thermo Scientific Chemicals

Catalog No. AC219230250
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219230250 25 g Plastic bottle
AC219235000 500 g Plastic bottle
AC219230025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC219230250 Supplier Thermo Scientific Chemicals Supplier No. 219230250

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 7789-29-9
Molecular Formula HF2K
Molecular Weight (g/mol) 78.1
MDL Number MFCD00011403
Synonym Potassium bifluoride

Specifications

Melting Point 239.0°C
Color White to Gray
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 99+%
Linear Formula KHF2
UN Number 1811
Fieser 16,283
Merck Index 15, 7731
Solubility Information Solubility in water: 39g/100mL (20°C). Other solubilities: soluble in diluted acids, insoluble in absolute alcohol, soluble in diluted alcohol
Formula Weight 78.1
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Potassium hydrogen fluoride, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 232-156-2
RTECSNumber TS6650000
TSCA TSCA

RUO – Research Use Only