CAS RN 7789-29-9
IUPAC Name:
potassium hydrogen difluoride
Synonyms:
potassium bifluoride
Molecular Weight (g/mol):
78.1
Molecular Formula:
F2HK
InChi Key:
VBKNTGMWIPUCRF-UHFFFAOYSA-M
SMILES:
[H+].[F-].[F-].[K+]
1
–
5
of
5
results
1
Potassium hydrogen fluoride, 99+% (metals basis), Thermo Scientific Chemicals
CAS: 7789-29-9 MDL Number: MFCD00011403 Synonym: Potassium bifluoride
2
Potassium hydrogen fluoride, 98%, Thermo Scientific Chemicals
CAS: 7789-29-9 Molecular Formula: KHF2 MDL Number: MFCD00011403 Synonym: Potassium bifluoride
3
Potassium hydrogen fluoride, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 7789-29-9 Molecular Formula: HF2K Molecular Weight (g/mol): 78.1 MDL Number: MFCD00011403 Synonym: Potassium bifluoride
4
Potassium Bifluoride (Potassium Hydrogen Fluoride), Reagents
CAS: 7789-29-9 Molecular Formula: F2HK Molecular Weight (g/mol): 78.10 InChI Key: VBKNTGMWIPUCRF-UHFFFAOYSA-M Synonym: Potassium Hydrogen Fluoride, Potassium Fluoride Hydrofluoride IUPAC Name: potassium hydrogen difluoride SMILES: [H+].[F-].[F-].[K+]
5