missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen fluoride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1303122

 View more versions of this product

Catalog No. AAA1303122


Only null left
Explore available promotions

Description

Description

Potassium hydrogen fluoride is used as an abrasive, as a pretreatment of metals before they are further processed, as a wood preservative and as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen fluoride is used as an abrasive, as a pretreatment of metals before they are further processed, as a wood preservative and as a chemical intermediate.

Solubility
Soluble in water (392 g/L). Also soluble in ethanol.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
Specifications

Specifications

Potassium hydrogen fluoride
7789-29-9
100 g
MFCD00011403
14,7610
Soluble in water (392g/L). Also soluble in ethanol.
98%
Hygroscopic
239°C (decomposition)
1
KHF2
UN1811
Potassium bifluoride
78.1
Odorless
2.37

Chemical Identifiers

7789-29-9
KHF2
MFCD00011403
Potassium bifluoride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314
Toxic if swallowed.
Causes severe skin burns and eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POTASSIUM HYDROGENDIFLUORIDE SOLID

EINECSNumber : 232-156-2

RTECSNumber : TS6650000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only