missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Potassium hydride, 30% w/w in mineral oil

Catalog No. AAL13266NB
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13266NB 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL13266NB Supplier Thermo Scientific™ Supplier No. L13266NB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7693-26-7 | HK | 40.11 g/mol

Potassium hydride is a precursor in the production of various bases by deprotonation, highly-hindered bases potassium triethylmethoxide and potassium 3-aminopropylamide It serves as a powerful base in organic synthesis of deprotonate carbonyl compounds and amines to form enolates and amides respectively.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 7693-26-7
Molecular Formula HK
Molecular Weight (g/mol) 40.11
MDL Number MFCD00011357
InChI Key OCFVSFVLVRNXFJ-UHFFFAOYSA-N
IUPAC Name potassium hydride
SMILES [H-].[K+]

Specifications

Density 1.45
Quantity 5 g
Flash Point 160°(320°F)
Form Liquid
UN Number UN1409
Fieser 1,935 12,407 13,257 14,265 17,290 18,299 19,276 21,360
Formula Weight 40.11
ShelfLife 5 Years
EINECSNumber 231-704-8