accessibility menu, dialog, popup

UserName

CAS RN 7693-26-7

IUPAC Name: potassium hydride
Synonyms: potassium hydride
Molecular Weight (g/mol): 40.11
Molecular Formula: HK
InChi Key: OCFVSFVLVRNXFJ-UHFFFAOYSA-N
SMILES: [H-].[K+]
Quantity 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results