missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexahydroxyplatinate(IV), Premion™, 99.95% (metals basis), Pt 51.5% min

Catalog No. AA4101403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4101403 1 g
AA4101406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4101403 Supplier Thermo Scientific Chemicals Supplier No. 04101403
Only null left

CAS: 12285-90-4 | H12K2O6Pt+2 | 381.371 g/mol

Potassium hexahydroxyplatinate(IV) is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12285-90-4
Molecular Formula H12K2O6Pt+2
Molecular Weight (g/mol) 381.371
MDL Number MFCD00064671
InChI Key LTROBPQZKJGHPB-UHFFFAOYSA-N
Synonym potassium hexahydroxoplatinate, dipotassium platinate, potassium hexahydroxyplatinate iv, platinum potassium hydroxide, h6k2o6pt, potassium hexahydroxoplatinate iv, potassium hexahydroxyplatinum iv, potassium hexahydroxyplatinate, dipotassium hexahydroxyplatinumdiuide, premion metalsbasis ,ptmin
PubChem CID 16717651
IUPAC Name dipotassium;platinum;hexahydrate
SMILES O.O.O.O.O.O.[K+].[K+].[Pt]

Specifications

Physical Form Powder
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 375.34
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Pt 51.5% min.
Assay (metals basis)
Chemical Name or Material Potassium hexahydroxyplatinate(IV)
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 235-554-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only