Learn More
Potassium hexafluorotitanate, 97%
Description
Potassium hexafluorotitanate is used as analytical reagent, alloying additive, flame retardant and flux agent for casting. It is also used in the manufacture of titanium acid and aluminum alloys containing titanium. It is also employed in metal surface treatment and finishing, detergent applications and dental impression molding. Further, it is used as a catalyst for polypropylene synthesis. In addition to this, it is used in the extraction-refining-processing of metals, electrical-electronics engineering, textile processing and in ceramics-glass industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium hexafluorotitanate is used as analytical reagent, alloying additive, flame retardant and flux agent for casting. It is also used in the manufacture of titanium acid and aluminum alloys containing titanium. It is also employed in metal surface treatment and finishing, detergent applications and dental impression molding. Further, it is used as a catalyst for polypropylene synthesis. In addition to this, it is used in the extraction-refining-processing of metals, electrical-electronics engineering, textile processing and in ceramics-glass industries.
Solubility
Soluble in hot water. Slightly soluble in cold water and inorganic acid. Insoluble in ammonia.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Boiling Point | 235°C to 237°C |
| Quantity | 2500 g |
| Linear Formula | K2TiF6 |
| UN Number | UN3288 |
| Solubility Information | Soluble in hot water. Slightly soluble in cold water and inorganic acid. Insoluble in ammonia. |
| Formula Weight | 240.07 |
| Percent Purity | 97% |
| Chemical Name or Material | Potassium hexafluorotitanate |
Safety and Handling
| Hazard Category | H302-H318 |
| Hazard Statement | GHS H Statement H318-H302-H317-H335-H315 Causes serious eye damage. Harmful if swallowed. May cause an allergic skin reaction. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S. |
| EINECSNumber | 240-969-9 |
| RTECSNumber | TT1575000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only