accessibility menu, dialog, popup

UserName

CAS RN 16919-27-0

FFFFFFKKTi

CAS RN 16919-27-0

IUPAC Name: titanium(4+) dipotassium hexafluoride
Synonyms: titanium(4+) dipotassium hexafluoride
Molecular Weight (g/mol): 240.05
Molecular Formula: F6K2Ti
InChi Key: RXCBCUJUGULOGC-UHFFFAOYSA-H
SMILES: [F-].[F-].[F-].[F-].[F-].[F-].[K+].[K+].[Ti+4]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results