missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluorotitanate, 97%

Catalog No. AAA1091522
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1091522 100 g
AAA1091536 500 g
AAA109150E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1091522 Supplier Thermo Scientific Chemicals Supplier No. A1091522
Only null left

CAS: 16919-27-0 | F6K2Ti

Potassium hexafluorotitanate is used as analytical reagent, alloying additive, flame retardant and flux agent for casting. It is also used in the manufacture of titanium acid and aluminum alloys containing titanium. It is also employed in metal surface treatment and finishing, detergent applications and dental impression molding. Further, it is used as a catalyst for polypropylene synthesis. In addition to this, it is used in the extraction-refining-processing of metals, electrical-electronics engineering, textile processing and in ceramics-glass industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hexafluorotitanate is used as analytical reagent, alloying additive, flame retardant and flux agent for casting. It is also used in the manufacture of titanium acid and aluminum alloys containing titanium. It is also employed in metal surface treatment and finishing, detergent applications and dental impression molding. Further, it is used as a catalyst for polypropylene synthesis. In addition to this, it is used in the extraction-refining-processing of metals, electrical-electronics engineering, textile processing and in ceramics-glass industries.

Solubility
Soluble in hot water. Slightly soluble in cold water and inorganic acid. Insoluble in ammonia.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 16919-27-0
Molecular Formula F6K2Ti
MDL Number MFCD00011380

Specifications

Boiling Point 235°C to 237°C
Quantity 100 g
Linear Formula K2TiF6
UN Number UN3288
Solubility Information Soluble in hot water. Slightly soluble in cold water and inorganic acid. Insoluble in ammonia.
Formula Weight 240.07
Percent Purity 97%
Chemical Name or Material Potassium hexafluorotitanate
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318-H302-H317-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 240-969-9
RTECSNumber TT1575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only