missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexacyanoferrate(II) trihydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™

Catalog No. 60045931
Change view
Click to view available options
Quantity:
500 g
6 x 500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60045931 6 x 500 g
6002335 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045931 Supplier Honeywell Chemicals Supplier No. 312546X500G
Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Quantity 6 x 500 g
Linear Formula K4Fe(CN)6 · 3H2O
UN Number NONH for all modes of transport
Formula Weight 422.39
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Chemical Name or Material Potassium Hexacyanoferrate(Ii) Trihydrate, Reag. Ph. Eur. reag. ISO
Hazard Statement P273
Precautionary Statement H412
EINECSNumber 237-722-2