missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexacyanoferrate(II) trihydrate, 98+%

Catalog No. AAA1573622
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1573622 100 g
AAA1573636 500 g
AAA157360E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1573622 Supplier Thermo Scientific Chemicals Supplier No. A1573622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14459-95-1 | C6H6FeK4N6O3 | 422.39 g/mol

Potassium hexacyanoferrate(II) trihydrate is widely used in anticaking agent for both road and table salt. It is involved in the purification of tin and separation of copper from molybdenum ores. It plays a vital role in the in production of wine and citric acid. In analytical chemistry, it is used to determine the concentration of potassium permanganate. It is in combination with potassium ferricyanide and phosphate buffered solution acts as buffer for beta-galatosidase. Further, it is used as fertilizers for plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hexacyanoferrate(II) trihydrate is widely used in anticaking agent for both road and table salt. It is involved in the purification of tin and separation of copper from molybdenum ores. It plays a vital role in the in production of wine and citric acid. In analytical chemistry, it is used to determine the concentration of potassium permanganate. It is in combination with potassium ferricyanide and phosphate buffered solution acts as buffer for beta-galatosidase. Further, it is used as fertilizers for plants.

Solubility
Soluble in water. Insoluble in ethanol and ether.

Notes
Begins to lose water at 600C, anhydrous at 1000C. Incompatible with oxidizing agents and strong acids.

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
Synonym Potassium ferrocyanide trihydrate
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point 69°C to 71°C
pH 9.5
Quantity 100 g
Odor Odorless
Density 1.85
Merck Index 14,7631
Solubility Information Soluble in water. Insoluble in ethanol and ether.
Formula Weight 422.39 (368.36 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Potassium hexacyanoferrate(II) trihydrate
EINECSNumber 237-722-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only