missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexachloroplatinate(IV), 99%+, MilliporeSigma™

Catalog No. p-4939062
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Fibre case
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1192380001 1 g Glass bottle
M1192380005 5 g Fibre case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1192380001 Supplier Technidata Supplier No. 1.19238.0001
Only null left

Chemical Identifiers

CAS 16921-30-5
Molecular Formula Cl6K2Pt
Molecular Weight (g/mol) 485.98
MDL Number MFCD00011389
InChI Key DPAIVKJGTXERIM-UHFFFAOYSA-H
Synonym potassium hexachloroplatinate iv, potassium platinic chloride, potassium hexachloroplatinate, dipotassium platinum hexachloride, platinic potassium chloride, dipotassium hexachloroplatinate, potassium platinum chloride, dipotassium hexachloroplatinate 2-, ccris 7824, platinum potassium chloride
PubChem CID 61856
IUPAC Name dipotassium hexachloroplatinumdiuide
SMILES [K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Physical Form Solid
Quantity 1 g
Assay Percent Range 99%+
UN Number UN 2923
Formula Weight 486.01
Odor Odorless
Packaging Glass bottle
Appearance Yellow-orange solid
Chemical Name or Material Potassium Hexachloroplatinate(IV)
Hazard Category H301: Toxic if swallowed.
H318: Causes serious eye damage.
H334: May cause allergy or asthma symptoms or breathing difficulties if inhaled.
H317: May cause an allergic skin reaction.
DOTInformation DOT Class: 8 (6.1), Packaging Group: III
EINECSNumber 240-979-3