missing translation for 'onlineSavingsMsg'
Learn More

Potassium Gluconate 99.0+%, TCI America™

Catalog No. G0040500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0040500G 500 g
G004025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0040500G Supplier TCI America Supplier No. G0040500G

Chemical Identifiers

CAS 299-27-4
Molecular Formula C6H11KO7
Molecular Weight (g/mol) 234.245
MDL Number MFCD00064211
InChI Key HLCFGWHYROZGBI-JJKGCWMISA-M
Synonym potassium gluconate, potassium d-gluconate, gluconic acid potassium salt, potassuril, kaon elixir, potassium 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate, katorin, potalium, potasoral, sirokal
PubChem CID 16760467
IUPAC Name potassium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[K+]

Specifications

Color White-Yellow
Physical Form Crystalline Powder
Quantity 500 g
Formula Weight 234.25
Percent Purity ≥99.0% (T)
Chemical Name or Material Potassium Gluconate
RTECSNumber LZ5230000
TSCA Yes