missing translation for 'onlineSavingsMsg'
Learn More

Potassium gluconate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013847
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013847 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013847 Supplier Merck Emd Millipore Supplier No. PHR11301G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Gluconate is a potassium salt of gluconic acid. It is a commonly administered nutrient supplement form of potassium.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 299-27-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C6H11 KO7
MDL Number MFCD00064211
Melting Point 183°C
Synonym Potassium D-gluconate; 2,3,4,5,6-Pentahydroxycaproic acid potassium salt; D-Gluconic acid potassium salt
Type Certified Reference Material
Quantity 1 g
Formula Weight 234.25
Grade Certified Reference Material
Show More Show Less