missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Gluconate, Anhydrous, USP, 97-103%, Spectrum™ Chemical
Description
Spectrum™ Chemical Potassium Gluconate, Anhydrous, USP is a mineral supplement used to treat or prevent low amounts of potassium in the blood. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystals |
| Quantity | 50 kg |
| Percent Purity | 97 to 103% |
| Grade | USP |
| Loss on Drying | 0.03 |
| Packaging | Fiber Drum |
| Chemical Name or Material | Potassium Gluconate |