missing translation for 'onlineSavingsMsg'
Learn More

Potassium Gluconate, Anhydrous, USP, 97-103%, Spectrum™ Chemical

Catalog No. 18605576
Change view
Click to view available options
Quantity:
500 g
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605576 500 g Poly Bottle
18605578 12 kg Poly Pail
18605579 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18605576 Supplier Spectrum Chemical Mfg Cor Supplier No. PO178500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

PO178, 299-27-4, C6H11KO7

Spectrum™ Chemical Potassium Gluconate, Anhydrous, USP is a mineral supplement used to treat or prevent low amounts of potassium in the blood. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 299-27-4
Molecular Formula C6H11KO7
Molecular Weight (g/mol) 234.25
InChI Key HLCFGWHYROZGBI-JJKGCWMISA-M
IUPAC Name potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES [K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 97 to 103%
Grade USP
Loss on Drying 0.03
Packaging Poly Bottle