missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, Anhydrous (White Powder/Certified), Fisher Chemical™

$416.26 - $1003.68

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
IUPAC Name potassium fluoride
SMILES [F-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
O6113500
SDS View Documents Promotion Details
Thermo Fisher Scientific
O6113500
500 g Poly Bottle
Each for $416.26
Case of 6 Each for $2,497.56
Only null left
 
O61131
SDS View Documents Promotion Details
Thermo Fisher Scientific
O61131
1 kg Poly Bottle
Each for $1,003.68
Case of 4 Each for $4,014.72
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7789-23-3
58.10
NROKBHXJSPEDAR-UHFFFAOYSA-M
522689
potassium fluoride
FK
MFCD00011398
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
CHEBI:66872
[F-].[K+]

Specifications

1505°C
7789-23-3
8 to 9
500 g
FK
MFCD00011398
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride
522689
58.09
Certified
0.5% max.
2.48g/cm³
860°C
White
Solid
≥98.5 %
KF
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
[F-].[K+]
58.10
CHEBI:66872
≥98.5%
Pass Test
Poly Bottle
Potassium Fluoride, Anhydrous
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 6.1, : Poison

SDS
missing translation for 'documents'

missing translation for 'documents'