missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, Anhydrous, Purified, 99%, Spectrum™ Chemical

Catalog No. 18612643
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612643 45 kg Poly Bottle
18612638 25 g Poly Bottle
18612639 125 g Poly Bottle
18612640 500 g Poly Bottle
18612641 2.5 kg Poly Bottle
18612642 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18612643 Supplier Spectrum Chemical Mfg Cor Supplier No. P162145KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
IUPAC Name potassium fluoride
SMILES [F-].[K+]

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 45 kg
Percent Purity 99%
Grade Purified
Packaging Poly Bottle
Chemical Name or Material Potassium Fluoride, Anhydrous

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.