missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, Anhydrous, Purified, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P162112KG

 View more versions of this product

Catalog No. 18612642


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7789-23-3
12 kg
MFCD00011398
[F-].[K+]
58.10
Purified
1
FK
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride
99%
Fiber Drum

Chemical Identifiers

7789-23-3
58.10
NROKBHXJSPEDAR-UHFFFAOYSA-M
[F-].[K+]
FK
MFCD00011398
potassium fluoride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.