missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride, ACS, 99% min, Thermo Scientific Chemicals

Catalog No. AA4221622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4221622 100 g
AA4221636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4221622 Supplier Thermo Scientific Chemicals Supplier No. 04221622
Only null left

Potassium fluoride is used in metal finishing, batteries, coatings and photographic chemicals. It is utilized for the study of ion-specific swelling and de-swelling of ampholytic polymer gels as well as in the measurement of electronic polarizabilities of ions in polymers of alkali halides. It finds application in the electronic industry as a metal surface treatment product. It is used as a preservative, a food additive, a catalyzer and a water absorbing agent. In the Finkelstein reaction, it is actively involved in the conversion of chlorocarbons to fluorocarbons using polar solvents like dimethyl formamide and ethylene glycol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
SMILES [F-].[K+]

Specifications

Boiling Point 1505°C
Melting Point 859°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99% min
Linear Formula KF
UN Number UN1812
Beilstein 3902818
Merck Index 14,7632
Solubility Information Solubility in water at 21° (96g/100mL); Freely soluble in boiling water; Soluble in aqueous HF, liquid NH3; Insoluble in anhydrous alcohol
IUPAC Name potassium;fluoride
Formula Weight 58.1
Percent Purity ≤99%
Grade ACS
Odor Odorless
Refractive Index 1.363
Sensitivity Hygroscopic
Density 2.48 g/mL
Chemical Name or Material Potassium fluoride
Show More Show Less
Hazard Category H301+H311+H331-H318
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: POTASSIUM FLUORIDE, SOLID
EINECSNumber 232-151-5
RTECSNumber TT0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only