missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

$87.10 - $579.34

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
SMILES [F-].[K+]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC201350250
SDS View Documents
Thermo Scientific Chemicals
201350250
25 g Plastic bottle
Each for $87.10
Only null left
 
AC201352500
SDS View Documents
Thermo Scientific Chemicals
201352500
250 g Plastic bottle
Each for $218.37
Only null left
 
AC201355000
SDS View Documents
Thermo Scientific Chemicals
201355000
500 g Plastic bottle
Each for $357.07
Case of 6 Each for $0.00
Only null left
 
AC201350010
SDS View Documents
Thermo Scientific Chemicals
201350010
1 kg Plastic bottle
Each for $579.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Chemical Identifiers

7789-23-3
58.10
NROKBHXJSPEDAR-UHFFFAOYSA-M
522689
[F-].[K+]
FK
MFCD00011398
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
CHEBI:66872

Specifications

1505°C
7789-23-3
Crystalline Powder, Crystals and/or Chunks
99%
KF
01,933; 02,346; 05,555; 06,481; 08,410; 10,325; 11,434; 12,406; 13,256; 15,167; 16,282
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium;fluoride
522689
58.09
Extra Pure
Plastic bottle
Potassium fluoride, 99%
860°C
White
25 g
FK
MFCD00011398
15, 7753
Solubility in water: 920g/L (18°C). Other solubilities: soluble in hydrofluoric acid,insoluble in alcohol unless water is present
[F-].[K+]
58.10
CHEBI:66872
99%
Pass Test
2.48
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air

GHS Signal Word: Danger

EINECSNumber : 232-151-5

RTECSNumber : TT0700000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only