missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog No. AA1098004
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1098004 2 g
AA1098009 10 g
AA1098018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1098004 Supplier Thermo Scientific Chemicals Supplier No. 01098004
Only null left

Potassium fluoride is used in organic chemistry as a fluorinating agent to replace other halide ions like chorine, bromine etc. It is used as a catalyst in the manufacture of polyurethanes. Further, it is widely used for etching of glass, in disinfectant formulations and manufacture of optical glass. It is used in metal finishing, batteries, coatings and photographic chemicals. It is utilized for the study of ion-specific swelling and de-swelling of ampholytic polymer gels as well as in the measurement of electronic polarizabilities of ions in polymers of alkali halides. It finds application in the electronic industry as a metal surface treatment product. It is used as a preservative, a food additive, a catalyzer and a water absorbing agent. In the Finkelstein reaction, it is actively involved in the conversion of chlorocarbons to fluorocarbons using polar solvents like dimethyl formamide and ethylene glycol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
SMILES [F-].[K+]

Specifications

Boiling Point 1505°C
Melting Point 859°C
Physical Form Crystalline
Quantity 2 g
Assay Percent Range 99.99% (metals basis)
Beilstein 3902818
Solubility Information Freely soluble in water. Insoluble in alcohol.
IUPAC Name potassium fluoride
Formula Weight 58.1
Refractive Index 1.363
Sensitivity Hygroscopic
Density 2.48 g/mL
Chemical Name or Material Potassium fluoride
Show More Show Less
Hazard Category H301+H311+H331-H318
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 232-151-5
RTECSNumber TT0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only