missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Ferrocyanide Trihydrate ACS MP Biomedicals
$96.05 - $96.05
Chemical Identifiers
| CAS | 14459-95-1 |
|---|---|
| Molecular Formula | C6H6FeK4N6O3 |
| Molecular Weight (g/mol) | 422.39 |
| MDL Number | MFCD00167023 |
| InChI Key | NLYKFUFIUWPPIZ-UHFFFAOYSA-N |
| Synonym | Potassium Hexacyanoferate II |
| IUPAC Name | tetrapotassium hexacyanoironbis(ylium) trihydrate |
| SMILES | O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15256080
|
MP Biomedicals Inc
0215256080 |
100 g |
Each for $96.05
|
|
|||||
Chemical Identifiers
| 14459-95-1 | |
| 422.39 | |
| NLYKFUFIUWPPIZ-UHFFFAOYSA-N | |
| tetrapotassium hexacyanoironbis(ylium) trihydrate |
| C6H6FeK4N6O3 | |
| MFCD00167023 | |
| Potassium Hexacyanoferate II | |
| O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N |
Specifications
| 14459-95-1 | |
| C6H6FeK4N6O3 | |
| Potassium Hexacyanoferate II | |
| O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N | |
| 422.39 | |
| ACS | |
| Potassium Ferrocyanide, Trihydrate |
| 100 g | |
| MFCD00167023 | |
| NLYKFUFIUWPPIZ-UHFFFAOYSA-N | |
| tetrapotassium hexacyanoironbis(ylium) trihydrate | |
| 422.39 | |
| 7.87 g/cm3 at 25 °C |