missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Ferrocyanide Trihydrate ACS MP Biomedicals
Specifications
Specifications
| Physical Form | Crystalline Powder |
| Quantity | 100 g |
| Formula Weight | 422.39 |
| Percent Purity | 101.73% |
| Grade | ACS |
| Density | 7.87 g/cm3 at 25 °C |
| Chemical Name or Material | Potassium Ferrocyanide, Trihydrate |