missing translation for 'onlineSavingsMsg'
Learn More

Potassium ferrocyanide trihydrate, 98.5%, ACS reagent

Catalog No. p-43118
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424130050 5 g Glass bottle
AC424135000 500 g Plastic bottle
AC424130025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC424130050 Supplier Thermo Scientific Chemicals Supplier No. 424130050
Only null left

CAS: 14459-95-1 | C6H6FeK4N6O3 | 422.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
Synonym Potassium hexacyanoferrate(II)
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point 70.0°C
Color Yellow
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 98.50%
Linear Formula K4Fe(CN)6·3H2O
Merck Index 15, 7752
Solubility Information Solubility in water: 270g/L (12°C). Other solubilities: insoluble in alcohol
Formula Weight 422.4
Percent Purity 98.5%
Grade ACS Reagent
Packaging Glass bottle
Specific Gravity 1.85
Density 1.8500g/mL
Chemical Name or Material Potassium ferrocyanide trihydrate
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only