missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferricyanide Solution, 1%, Fisher Scientific™

$159.33 - $159.33

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
SP9401-500
View Documents
Ricca Chemical Company
SP9401500
500 mL Natural Poly Bottle
Each for $159.33
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13746-66-2
329.25
QMTKJUMXUDIUAQ-UHFFFAOYSA-N
26250
[K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
C6FeK3N6
MFCD00011392
potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
tripotassium hexacyanoirontris(ylium)

Specifications

13746-66-2,7732-18-5
100.0,1.01
MFCD00011392
QMTKJUMXUDIUAQ-UHFFFAOYSA-N
tripotassium hexacyanoirontris(ylium)
26250
Laboratory
For AOAC Method 953.08
Potassium Ferricyanide Solution, 1%
97.0,0.97
C6FeK3N6
potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
[K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
329.25
1%
Natural Poly Bottle
500 mL
Videos
Safety and Handling

Safety and Handling

SDS
Documents

Documents