missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferricyanide, Certified, 10.0% (w/v) ±0.5%, LabChem™

Catalog No. p-5388170
Change view
Click to view available options
Quantity:
500 mL
1 L
Catalog No. Quantity
LC190501 500 mL
LC190502 1 L
2 options

Catalog No. LC190501

Supplier: LabChem LC190501

Only null left

Chemical Identifiers

CAS 13746-66-2 , 7732-18-5
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Color Orange-Yellow
CAS Max % 90.48, 9.52
Physical Form Liquid
Quantity 500 mL
Linear Formula K3Fe(CN)6
Solubility Information Soluble in water
Formula Weight 329.26
Percent Purity 10.0% ±0.5% (w/v)
Grade Certified
Identification Passes Test
Packaging Amber Poly
Specific Gravity 1.05g/mL
Decomposition Information Hydrogen cyanide; Carbon monoxide; Carbon dioxide
Density 1.05g/mL
Chemical Name or Material Potassium Ferricyanide
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature