missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Ethylxanthate 95.0+%, TCI America™
Supplier: TCI America E0194100G
Specifications
| Potassium Ethylxanthate | |
| 210°C | |
| C3H5KOS2 | |
| 100 g | |
| JCBJVAJGLKENNC-UHFFFAOYSA-M | |
| potassium (ethoxymethanethioyl)sulfanide | |
| 2735045 | |
| ≥95.0% (T) |
| 140-89-6 | |
| White-Yellow | |
| MFCD00004931 | |
| potassium ethylxanthate, potassium ethyl xanthate, ethylxanthic acid potassium salt, potassium xanthogenate, potassium xanthate, potassium o-ethyl carbonodithioate, potassium ethyl xanthogenate, potassium ethylxanthogenate, potassium o-ethyl dithiocarbonate, z 3 pesticide | |
| [K+].CCOC([S-])=S | |
| 160.29 | |
| 160.29 | |
| Crystalline Powder |
Chemical Identifiers
| 140-89-6 | |
| 160.29 | |
| JCBJVAJGLKENNC-UHFFFAOYSA-M | |
| 2735045 | |
| [K+].CCOC([S-])=S |
| C3H5KOS2 | |
| MFCD00004931 | |
| potassium ethylxanthate, potassium ethyl xanthate, ethylxanthic acid potassium salt, potassium xanthogenate, potassium xanthate, potassium o-ethyl carbonodithioate, potassium ethyl xanthogenate, potassium ethylxanthogenate, potassium o-ethyl dithiocarbonate, z 3 pesticide | |
| potassium (ethoxymethanethioyl)sulfanide |
Safety and Handling
EINECSNumber : (2)-1787
RTECSNumber : FG1575000
TSCA : Yes