missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ethylxanthate 95.0+%, TCI America™

Catalog No. p-7128890
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E019425G 25 g
E0194100G 100 g
E0194500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E019425G Supplier TCI America Supplier No. E019425G

Chemical Identifiers

CAS 140-89-6
Molecular Formula C3H5KOS2
Molecular Weight (g/mol) 160.29
MDL Number MFCD00004931
InChI Key JCBJVAJGLKENNC-UHFFFAOYSA-M
Synonym potassium ethylxanthate, potassium ethyl xanthate, ethylxanthic acid potassium salt, potassium xanthogenate, potassium xanthate, potassium o-ethyl carbonodithioate, potassium ethyl xanthogenate, potassium ethylxanthogenate, potassium o-ethyl dithiocarbonate, z 3 pesticide
PubChem CID 2735045
IUPAC Name potassium (ethoxymethanethioyl)sulfanide
SMILES [K+].CCOC([S-])=S

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 25 g
Formula Weight 160.29
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Ethylxanthate
EINECSNumber (2)-1787
RTECSNumber FG1575000
TSCA Yes