missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Disulfite, puriss. p.a., Honeywell Fluka™
Puriss. p.a., powder,≥96% (iodometric)
Supplier: Honeywell Chemicals 3126825KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Potassium Disulfite | |
| Powder | |
| K2O5S2 | |
| MFCD00167605 | |
| potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5 | |
| [K+].[K+].[O-]S(=O)S([O-])(=O)=O | |
| 222.31 | |
| 222.32g/mol | |
| puriss. p.a. |
| 16731-55-8 | |
| 2.5 kg | |
| K2S2O5 | |
| NONH for all modes of transport | |
| RWPGFSMJFRPDDP-UHFFFAOYSA-L | |
| dipotassium sulfinatosulfonate | |
| 28019 | |
| ≥96% (iodometric) |
Chemical Identifiers
| 16731-55-8 | |
| 222.31 | |
| RWPGFSMJFRPDDP-UHFFFAOYSA-L | |
| 28019 | |
| [K+].[K+].[O-]S(=O)S([O-])(=O)=O |
| K2O5S2 | |
| MFCD00167605 | |
| potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5 | |
| dipotassium sulfinatosulfonate |
Safety and Handling
P280-P305 + P351 + P338
H318
EINECSNumber : 240-795-3
RTECSNumber : TT4920000