missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium dihydrogen phosphate, Anhydrous, 99.995% Suprapur™, MilliporeSigma™
$540.14 - $1831.89
Chemical Identifiers
| CAS | 7778-77-0 |
|---|---|
| Molecular Formula | H2KO4P |
| Molecular Weight (g/mol) | 136.08 |
| MDL Number | MFCD00011401 MFCD00147253 |
| InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
| Synonym | potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate |
| PubChem CID | 516951 |
| ChEBI | CHEBI:63036 |
| IUPAC Name | potassium dihydrogen phosphate |
| SMILES | [K+].OP(O)([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1051080050
|
Technidata
1051080050 |
50 g | Plastic Bottle |
Each for $540.14
|
|
||||
|
M1051080500
|
Technidata
1051080500 |
500 g | Plastic Bottle |
Each for $1,831.89
|
|
||||
Chemical Identifiers
| 7778-77-0 | |
| 136.08 | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| 516951 | |
| potassium dihydrogen phosphate |
| H2KO4P | |
| MFCD00011401 MFCD00147253 | |
| potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
| CHEBI:63036 | |
| [K+].OP(O)([O-])=O |
Specifications
| Colorless | |
| Solid | |
| 99.995% | |
| KH2PO4 | |
| potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
| [K+].OP(O)([O-])=O | |
| 136.08 | |
| CHEBI:63036 | |
| Suprapur | |
| 2.34g/cm3 (20°C); Bulk Density: 1200 kg/m3 |
| 7778-77-0 | |
| 50 g | |
| H2KO4P | |
| MFCD00011401 MFCD00147253 | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| potassium dihydrogen phosphate | |
| 516951 | |
| 136.8 | |
| Plastic Bottle | |
| Suprapur™ |
Safety and Handling
Recommended Storage : Store without restrictions