missing translation for 'onlineSavingsMsg'
Learn More

Potassium dichromate, MilliporeSigma™

Catalog No. p-5547646
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1048640500 500 g Plastic Bottle
M1048641000 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1048640500 Supplier Technidata Supplier No. 1.04864.0500
Only null left

For analysis EMSURE™ ACS, ISO, Reag. Ph Eur

Chemical Identifiers

CAS 7778-50-9
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Boiling Point >500°C
Quantity 500 g
Solubility Information Soluble in water: 130g/L at 10°C
Formula Weight 294.19
Odor Odorless
Packaging Plastic Bottle
Density 2.7g/cm3 at 20°C
Chemical Name or Material Potassium dichromate
Hazard Category H340: May cause genetic defects.
Hazard Statement H350: May cause cancer.
Precautionary Statement H360FD: May damage fertility. May damage the unborn child.
EINECSNumber 231-906-6
RTECSNumber HX7680000
Recommended Storage Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry. Do not store near combustible material. Keep locked up or in an area accessible only to qualified or authorized persons.