Learn More
Potassium Dichromate, Certified, 0.250N ±0.002N (0.0417M), LabChem™
Supplier: LabChem LC190106
Specifications
| Potassium Dichromate | |
| Orange | |
| Liquid | |
| Cr2K2O7 | |
| potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash | |
| KMUONIBRACKNSN-UHFFFAOYSA-N | |
| dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
| 24502 | |
| 294.18 | |
| 0.250N ±0.002N (0.0417M) | |
| Cubitainer™ | |
| Contains hexavalent Chromium |
| 7778-50-9,7732-18-5 | |
| 98.77,1.23 | |
| 10 L | |
| K2Cr2O7 | |
| Soluble in water | |
| [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] | |
| 294.182 | |
| CHEBI:53444 | |
| Certified | |
| Passes Test | |
| 1.01g/mL | |
| 1.01g/mL |
Chemical Identifiers
| 7778-50-9 | |
| 294.182 | |
| potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash | |
| CHEBI:53444 | |
| [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
| Cr2K2O7 | |
| KMUONIBRACKNSN-UHFFFAOYSA-N | |
| 24502 | |
| dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs (blood, kidneys, liver) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If experiencing respiratory symptoms: Call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature